Learn More
Electron Microscopy Sciences ProPlate™ Oncyte Avid 96 NC Round Pad, Glass Substrate
SDP
Catalog No. 5034345
Encompass_Preferred
Patented ProPlate technology for high throughput robotic processing of microarrays. Grace BioLabs developed bottomless SBS compliant microtiter plate superstructures for use with robotic processors. Features guaranteed flat SBS compliant microtiter plate format which enables robotic processing of assays. It is compatible with glass or plastic substrates and has a water-tight seal between superstructure and substrate. ONCYTE Nitrocellulose Coated Substrates, 6 mm Diameter for SBS Microtiter Plate, Glass Substrate.
Catalog No. 50-343-45 Supplier Electron Microscopy Sciences Supplier No. 6348520
Patented ProPlate technology for high throughput robotic processing of microarrays. Grace BioLabs developed bottomless SBS compliant microtiter plate superstructures for use with robotic processors. Features guaranteed flat SBS compliant microtiter plate format which enables robotic processing of assays. It is compatible with glass or plastic substrates and has a water-tight seal between superstructure and substrate. ONCYTE Nitrocellulose Coated Substrates, 6 mm Diameter for SBS Microtiter Plate, Glass Substrate.