Learn More
Electron Microscopy Sciences Oncyte Slide Avid, 4 well, 15 x 15 mm I.D.
SDP
Catalog No. 5027526
Encompass_Preferred
ONCYTE is a three-dimensional microarray comprised of a combination of nitrocellulose polymer and proprietary chemistry that provides increased protein affinity and binding. It also provides enhanced fluorescent chemiluminescent, radiographic, colormetric and visual signal compared to glass or other coated glass surfaces. AVID for the highest possible protein binding for colometric and infrared.
Wells: 4
I.D.: 15 x 15 mm
Catalog No. 50-275-26 Supplier Electron Microscopy Sciences Supplier No. 7033561A
ONCYTE is a three-dimensional microarray comprised of a combination of nitrocellulose polymer and proprietary chemistry that provides increased protein affinity and binding. It also provides enhanced fluorescent chemiluminescent, radiographic, colormetric and visual signal compared to glass or other coated glass surfaces. AVID for the highest possible protein binding for colometric and infrared.
Wells: 4
I.D.: 15 x 15 mm