Learn More
Electron Microscopy Sciences Oncyte Microtiter Plates Avid, 96 round, 6 mm I.D.
SDP
Catalog No. 5027532
Encompass_Preferred
ONCYTE is a three-dimensional microarray comprised of a combination of nitrocellulose polymer and proprietary chemistry that provides increased protein affinity and binding. It also provides enhanced fluorescent chemiluminescent, radiographic, colormetric and visual signal compared to glass or other coated glass surfaces. AVID for Highest possible protein binding. SBS Compliant w/Glass Substrate.
Wells: 96
I.D.: 6 mm diameter
Catalog No. 50-275-32 Supplier Electron Microscopy Sciences Supplier No. 7033570A
ONCYTE is a three-dimensional microarray comprised of a combination of nitrocellulose polymer and proprietary chemistry that provides increased protein affinity and binding. It also provides enhanced fluorescent chemiluminescent, radiographic, colormetric and visual signal compared to glass or other coated glass surfaces. AVID for Highest possible protein binding. SBS Compliant w/Glass Substrate.
Wells: 96
I.D.: 6 mm diameter