Learn More
Invitrogen™ Avidin
Description
Reactivity: Avidin reacts with biotin.
Specifications
Specifications
| Conjugate | Unconjugated |
| Quantity | 10 mg |
| Content And Storage | Store in refrigerator (2–8°C). |
| Shipping Condition | Wet Ice |
| Form | Lyophilized |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.