Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Avidin, Alexa Fluor™ 488 conjugate

Catalog No. A21370
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21370 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A21370 Supplier Invitrogen™ Supplier No. A21370
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Avidin, Alexa Fluor™ 488 conjugate

Our avidin, Alexa Fluor™ 488 conjugate is a bright, photostable probe with excitation and emission characteristics similar to those of fluorescein. Avidin is a glycosylated biotin-binding protein that has a basic isoelectric point and is widely used to detect biotinylated probes.

TRUSTED_SUSTAINABILITY

Specifications

Conjugate Alexa Fluor 488
Quantity 1 mg
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Shipping Condition Room Temperature
Form Solid
Product Line Alexa Fluor
Label or Dye Alexa Fluor Dyes
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.