Learn More
Invitrogen™ Avidin/Biotin Blocking Kit
Description
This kit is used to block endogenous biotin or biotin-binding activity in tissue sections. The kit contains 18 mL each of Avidin and Biotin solutions.
Specifications
Specifications
| Quantity | 1 Kit |
| Content And Storage | Store in refrigerator (2–8°C). |
| Shipping Condition | Wet Ice |
| Product Type | Avidin/Biotin Blocking Kit |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.