Learn More
Abcam Avidin/Biotin Blocking Kit, 15ML
Catalog No. NC1902772
Encompass_Preferred
Avidin/biotin blocking kit blocks signals from endogenous avidin, biotin and biotin-binding proteins in tissues when used with biotin-based IHC detection. - Cited in over 35 publications - Ideal for tissues rich in biotin (e.g., liver, kidney) - Easy-to-use, quick protocol
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1902772 Supplier Abcam Supplier No. AB6421215ML
Avidin/biotin blocking kit blocks signals from endogenous avidin, biotin and biotin-binding proteins in tissues when used with biotin-based IHC detection. - Cited in over 35 publications - Ideal for tissues rich in biotin (e.g., liver, kidney) - Easy-to-use, quick protocol
The product is subject to the following: Abcam Restricted Use Statement