Learn More
Biocare Medical, LLC Avidin-Biotin Kit - 100 ml
SDP
Catalog No. 5082359
Encompass_Preferred
The Avidin and Biotin solutions are color-coded with a pH indicator. In most cases, endogenous biotin in tissue sections is masked by formalin fixation. However, if avidin-biotin IHC detection systems are used with frozen sections or tissues pretreated with a heat-induced, epitope retrieval method (HIER), an avidin-biotin blocking technique may be needed. It can also be used on an automated staining system.
Catalog No. 50-823-59 Supplier Biocare Medical, LLC Supplier No. AB972L
The Avidin and Biotin solutions are color-coded with a pH indicator. In most cases, endogenous biotin in tissue sections is masked by formalin fixation. However, if avidin-biotin IHC detection systems are used with frozen sections or tissues pretreated with a heat-induced, epitope retrieval method (HIER), an avidin-biotin blocking technique may be needed. It can also be used on an automated staining system.