Learn More
Biocare Medical, LLC Avidin-Biotin Kit - 500 ml
SDP
Catalog No. NC0194849
Encompass_Preferred
The Avidin and Biotin solutions are color-coded with a pH indicator. In most cases, endogenous biotin in tissue sections is masked by formalin fixation. However, if avidin-biotin IHC detection systems are used with frozen sections or tissues pretreated with a heat-induced, epitope retrieval method (HIER), an avidin-biotin blocking technique may be needed. It can also be used on an automated staining system.
Catalog No. NC0194849 Supplier Biocare Medical, LLC Supplier No. AB972M
The Avidin and Biotin solutions are color-coded with a pH indicator. In most cases, endogenous biotin in tissue sections is masked by formalin fixation. However, if avidin-biotin IHC detection systems are used with frozen sections or tissues pretreated with a heat-induced, epitope retrieval method (HIER), an avidin-biotin blocking technique may be needed. It can also be used on an automated staining system.