Learn More
Medchemexpress LLC Avidin, chicken egg white | 1405-69-2 | 99.9% | 1 MG
SDP
Catalog No. 5000426044
Encompass_Preferred
Avidin, chicken egg white is a glycoprotein derived from egg protein. It has excellent affinity with biotin and is often used in combination with biotin for immunoassays to detect the location of antigens in tissues.
  • Derived from egg protein
  • Exhibits excellent affinity with biotin
  • Suitable for immunoassays
  • Detects the location of antigens in tissues
Catalog No. 50-004-26044 Supplier Medchemexpress LLC Supplier No. HYNP0051MG
Avidin, chicken egg white is a glycoprotein derived from egg protein. It has excellent affinity with biotin and is often used in combination with biotin for immunoassays to detect the location of antigens in tissues.
  • Derived from egg protein
  • Exhibits excellent affinity with biotin
  • Suitable for immunoassays
  • Detects the location of antigens in tissues