Learn More
Vector Laboratories Avidin D 10 mg
SDP
Supplier:  Vector Laboratories A200010
Encompass
Avidin D is isolated from egg white by a specific affinity system which results in a homogeneous, ultrapure avidin with very low non-specific binding. Avidin D is recommended for use in solid-phase assays and many histochemical applications where sequential addition of reagents is employed.
Catalog No. NC9123216