Learn More
Vector Laboratories Avidin D, Horseradish Peroxidase (Av-HRP) Concentrate For ELISA and Blots 5 mg
SDP
Supplier:  Vector Laboratories A20145
Encompass
Vector LaboratoriesÆ enzyme-conjugated avidin and streptavidin products are produced with high specific activity enzymes in optimal ratios. Covalent linkages are chosen to provide stable, highly active conjugates. This horseradish peroxidase-labeled Avidin D (Av-HRP) is optimized for use in solid-phase assays such as ELISAs and blotting applications.
Catalog No. NC0738459