Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Avidin from Egg White

Catalog No. MP215004780 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215004780 100 mg
MP215004705 5 mg
MP215004710 10 mg
MP215004725 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP215004780 Supplier MP Biomedicals™ Supplier No. 0215004780
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Avidin from Egg White, 10-15 u/mg solid, White Crystalline Powder

  • Avidin is useful in a biotin-avidin system for labeling biomolecules for receptor studies, immunoassays and immunohistological methods.
  • Avidin egg white was used in an assay using functionalized xenon as a biosensor to detect biotin-avidin binding.
  • Egg white was used at 80 nmol.
  • Egg white avidin has been used as a cross-linker for biotin-conjugated antibodies.
  • The product has also been used as a blocking agent in ELISA applications.
  • Avidin is a basic glycoprotein isolated from raw egg white.
  • It exhibits high binding affinity for biotin and is capable of producing biotin deficiency in rats and chicks.
  • It also occurs in the white portion of eggs and the oviducts of birds and amphibia.
  • It is destroyed by cooking or irradiation.
  • One unit will bind 1 ug of D-biotin at pH 8.9.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Quantity 100 mg
Source Egg White
CAS 1405-69-2
Content And Storage -20°C
Form Crystalline
For Use With (Application) Labeling biomolecules for receptor studies, immunoassays and immunohistological methods
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.