Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ Avidin HRP

Catalog No. 5015377
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-153-77 500 μL
50-112-8941 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-153-77 Supplier Invitrogen™ Supplier No. 18410094
  • $127.00
    $123.65 / Each of 1

    Save $3.35

    3% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Commonly used as a second step for detection of biotinylated antibodies by ELISA and western blot. Avidin binds to biotin with high affinity. [Note: Sodium azide is an inhibitor of horseradish peroxidase enzymatic activity and should not be added to the AV-HRP diluent.]

Avidin binds to biotin with high affinity. Because sodium azide is an inhibitor of horseradish peroxidase enzymatic activity, it should not be added to the AV-HRP diluent.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 500 μL
Target Molecule Biotin
Form Liquid
Product Line eBioscience
Label or Dye HRP (Horseradish Peroxidase)
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.