Learn More
Invitrogen™ Avidin, NeutrAvidin™, Horseradish Peroxidase conjugate
Description
Our horseradish peroxidase (HRP) conjugate of NeutrAvidin™ biotin-binding protein - a form of avidin that has been processed to remove carbohydrate and to lower its isoelectric point - can substantially decrease background due to nonspecific binding. The method used to deglycosylate the avidin retains its specific binding.
Specifications
Specifications
| Conjugate | HRP |
| Quantity | 1 mg |
| Content And Storage | Store in freezer (-5 to -30°C). |
| Shipping Condition | Wet Ice |
| Product Line | NeutrAvidin |
| Label or Dye | HRP (Horseradish Peroxidase) |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.