Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Avidin, NeutrAvidin™, PE conjugate

Catalog No. a2660
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2660 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2660 Supplier Invitrogen™ Supplier No. A2660
Only null left
Add to Cart
Edge
Add to Cart

Avidin, NeutrAvidin™, PE conjugate

Our R-phycoerythrin (R-PE) conjugate of NeutrAvidin™ biotin-binding protein - a form of avidin that has been processed to remove carbohydrate and to lower its isoelectric point - can substantially decrease background due to nonspecific binding. The method used to deglycosylate the avidin retains its specific binding. This bright, fluorescent reagent is ideal for detecting biotinylated targets in flow cytometry applications.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL
Conjugate R-phycoerythrin
Quantity 1 mL
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Shipping Condition Wet Ice
Product Line NeutrAvidin
Product Type Avidin
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.