Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Avidin, NeutrAvidin™, Texas Red™ conjugate

Catalog No. A2665
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2665 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2665 Supplier Invitrogen™ Supplier No. A2665
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Avidin, NeutrAvidin™, Texas Red™ conjugate

Our Texas Red™ conjugate of NeutrAvidin™ biotin-binding protein - a form of avidin that has been processed to remove carbohydrate and to lower its isoelectric point - can substantially decrease background due to nonspecific binding. The method used to deglycosylate the avidin retains its specific binding. This bright, red-fluorescent reagent (absorption/emission maxima ∼595/615 is ideal for detecting biotinylated targets in fluorescence microscopy applications.

TRUSTED_SUSTAINABILITY

Specifications

Conjugate Texas Red
Quantity 1 mg
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Shipping Condition Room Temperature
Product Line NeutrAvidin, Texas Red
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.