Learn More
Sigma Aldrich Fine Chemicals Biosciences Avidin Peroxidase buffered
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A74192ML
Encompass
Avidin is homotetrameric protein (68kDa) obtained from egg whites and binds strongly to biotin. Thus avidin-biotin association has been utilized in immunoassays to detect the localization of antigens in tissues. The use of avidin-biotin immunoassay enhances the sensitivity of the technique and facilitates the detection of antigens in low quantities.
Catalog No. 50-290-5853