Learn More
Bioworld AVIDIN PROTEIN
Supplier:  Bioworld 220700012
Encompass_Preferred
Avidin binds with biotin making it a useful product in the diagnostic industry and has applications in purification of proteins.; Avidin is used for the targeting of solid tumors and for applications in vitro such as protein purification...
Catalog No. 50-253-886