Learn More
Bioworld AVIDIN SEPAROPOREAGAROSE) 6B
Supplier:  Bioworld 201810603
Encompass
Specifications: Ligand: Avidin (from egg white); Particle size (range): 50-175m; Molecular Weight range: 1x104 - 4x106; Matrix: Separopore 6B-CL (Crosslinked Agarose 6% Beads); pH stability: 4-9; Flow specifications: 70-140 cm/h; Supplie...
Catalog No. 50-197-818