Learn More
Abcam Avidin sulforhodamine 101 conjugate (Te x as Red Avidin equivalent), 5MG
Catalog No. 50996212
Encompass_Preferred
MW 67000 g/mol. Fluorescent conjugate useful for DNA and RNA microarray analyses and in detecting biotin-labeled antibodies. It can be employed with other fluorescein or coumarin conjugates to simultaneously localize two or three antigens in the same tissue section - excitation/emission maxima are well separated from those of fluorescein.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-212 Supplier Abcam Supplier No. AB2753085MG
MW 67000 g/mol. Fluorescent conjugate useful for DNA and RNA microarray analyses and in detecting biotin-labeled antibodies. It can be employed with other fluorescein or coumarin conjugates to simultaneously localize two or three antigens in the same tissue section - excitation/emission maxima are well separated from those of fluorescein.
The product is subject to the following: Abcam Restricted Use Statement