Learn More
Sigma Aldrich Fine Chemicals Biosciences Avidin Sulforhodamine 1015MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A23485MG
Encompass
Highly cationic glycoprotein that can selectively bind to a component in human and rodent mast cell granules in fixed-cell preparations and can be used to identify mast cells in normal and diseased human tissue without requiring a biotinylated probe.
Catalog No. 50-290-5197