Learn More
Invitrogen™ Avidin, Texas Red™ conjugate
Description
Our avidin, Texas Red™ conjugate is a bright, photostable probe with excitation and emission maxima ∼595/615. Avidin is a glycosylated biotin-binding protein that has a basic isoelectric point and is widely used to detect biotinylated probes.
Specifications
Specifications
| Conjugate | Texas Red |
| Quantity | 5 mg |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Shipping Condition | Room Temperature |
| Product Line | Texas Red |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.