Learn More
Aviva Systems Biology Corp AVIL Antibody - middle region (ARP32811_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32811_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AVIL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human AVIL. Alternate names: ADVIL, DKFZp779O1812, DOC6, FLJ12386, MGC133244, p92ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54778