Learn More
ABclonal Technology AVIL Rabbit pAb
SDP
Supplier:  ABclonal Technology A15785500UL
Encompass_Preferred
The protein encoded by this gene is a member of the gelsolin/villin family of actin regulatory proteins. This protein has structural similarity to villin. It binds actin and may play a role in the development of neuronal cells that form ganglia.
Catalog No. 50-001-08043