Learn More
Cayman Chemical Avilamycin A, Purity: ≥75%, CAS: 69787-79-7, 50 mg
SDP
Catalog No. 502938060
Encompass_Preferred
An antibiotic; active against veterinary isolates of C. perfringens (MICs = ≤0.06-0.5 mg/L) and B. hyodysenteriae (MICs = 12.5-100 μg/ml); dietary administration reduces mortality in a broiler cockerel model of C. perfringens infection at 15 and 30 ppm
Catalog No. 50-293-8060 Supplier Cayman Chemical Supplier No. 2913550 MG
An antibiotic; active against veterinary isolates of C. perfringens (MICs = ≤0.06-0.5 mg/L) and B. hyodysenteriae (MICs = 12.5-100 μg/ml); dietary administration reduces mortality in a broiler cockerel model of C. perfringens infection at 15 and 30 ppm