Learn More
Perkin Elmer US LLC Avio 200 Quartz Baffled Cyclonic Spray Chamber
Catalog No. 502112089
Encompass_Preferred
The Baffled Cyclonic Spray Chamber features a central transfer tube which acts as a secondary particle separator, helping to separate larger aerosol particles from the sample. This reduces solvent load in the plasma without compromising detection limits.
Catalog No. 50-211-2089 Supplier Perkin Elmer US LLC Supplier No. N0791841
The Baffled Cyclonic Spray Chamber features a central transfer tube which acts as a secondary particle separator, helping to separate larger aerosol particles from the sample. This reduces solvent load in the plasma without compromising detection limits.