Learn More
TARGETMOL CHEMICALS INC Aviptadil Acetate 5MG
SDP
Catalog No. 502880269
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Aviptadil Acetate (Vasoactive Intestinal Peptide acetate salt) induces pulmonary vasodilation and inhibits the proliferation of vascular SMCs and platelet aggregation. Avitadil acetate can be used for research on pulmonary fibrosis, pulmonary hypertension (PAH), and respiratory failure caused by SARS-CoV-2. Purity 100%
Catalog No. 50-288-0269 Supplier TARGETMOL CHEMICALS INC Supplier No. TP1016L5MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Aviptadil Acetate (Vasoactive Intestinal Peptide acetate salt) induces pulmonary vasodilation and inhibits the proliferation of vascular SMCs and platelet aggregation. Avitadil acetate can be used for research on pulmonary fibrosis, pulmonary hypertension (PAH), and respiratory failure caused by SARS-CoV-2. Purity 100%