Learn More
REPLIGEN CORPORATION AVIPure AAV9 Settled Resin, High Dynamic Binding Capacity, 25 mL
SDP
Catalog No. NC2380994
Encompass
The AVIPure - AAV9 25 mL Settled Resin is an advanced affinity chromatography resin specifically designed for the purification of adeno-associated virus (AAV9) vectors. It benefits customers by providing high dynamic binding capacity for efficient capture from clarified cell culture fluids, robust alkaline stability for multi-cycle usage, and compatibility with various buffers and solvents, setting it apart from traditional options in gene therapy manufacturing.
Catalog No. NC2380994 Supplier REPLIGEN CORPORATION Supplier No. 100AAV925
The AVIPure - AAV9 25 mL Settled Resin is an advanced affinity chromatography resin specifically designed for the purification of adeno-associated virus (AAV9) vectors. It benefits customers by providing high dynamic binding capacity for efficient capture from clarified cell culture fluids, robust alkaline stability for multi-cycle usage, and compatibility with various buffers and solvents, setting it apart from traditional options in gene therapy manufacturing.