Learn More
MSC National Chemical Laboratories Disinfectant Cleaner, Ready-To-Use, Citrus Lemon, 32 oz
Catalog No. 502867066
Encompass
This product is a general, ready-to-use cleaner and disinfectant designed for use on hard, non-porous environmental surfaces across various settings, including healthcare, commercial, and residential. It effectively disinfects and deodorizes surfaces prone to odors caused by microorganisms.
  • One-step disinfectant cleaner
  • Effective against MRSA, CA-MRSA, HIV-1, HBV, HCV, and Mycobacterium Tuberculosis (TB)
  • Non-acid formulation
  • Provides long-lasting odor control
  • Powerful disinfectant effective in the presence of organic soil (5% blood serum)
  • Liquid form in a 32 oz bottle
  • Features citrus and lemon scent
  • EPA registered formula based on quaternary ammonium chlorides
  • Ready-to-use solution with 0.2% VOC content
Catalog No. 50-286-7066 Supplier MSC Supplier No. 16979874
This product is a general, ready-to-use cleaner and disinfectant designed for use on hard, non-porous environmental surfaces across various settings, including healthcare, commercial, and residential. It effectively disinfects and deodorizes surfaces prone to odors caused by microorganisms.
  • One-step disinfectant cleaner
  • Effective against MRSA, CA-MRSA, HIV-1, HBV, HCV, and Mycobacterium Tuberculosis (TB)
  • Non-acid formulation
  • Provides long-lasting odor control
  • Powerful disinfectant effective in the presence of organic soil (5% blood serum)
  • Liquid form in a 32 oz bottle
  • Features citrus and lemon scent
  • EPA registered formula based on quaternary ammonium chlorides
  • Ready-to-use solution with 0.2% VOC content