Learn More
Selleck Chemical LLC Avitinib (AC0010) 5mg 1557267-42-1
SDP
Supplier:  Selleck Chemical LLC S87415MG
Encompass_Preferred
Avitinib (AC0010) is a pyrrolopyrimidine-based irreversible EGFR inhibitor that is mutation-selective with IC50 value of 0.18 nM against EGFR L858R/T790M double mutations, nearly 43-fold greater potency over wild-type EGFR (IC50 value, 7.68 nM). It has comparable anti-tumor activity and tolerated toxicity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8391