Learn More
TARGETMOL CHEMICALS INC AVITINIB MALEATE 10MG
SDP
Catalog No. 502576878
Encompass_Preferred
Also available in 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Avitinib maleate is a pyrrolopyrimidine-based irreversible epidermal growth factor receptor (EGFR) inhibitor. purity: 99%
Catalog No. 50-257-6878 Supplier TARGETMOL CHEMICALS INC Supplier No. T372110MG
Also available in 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Avitinib maleate is a pyrrolopyrimidine-based irreversible epidermal growth factor receptor (EGFR) inhibitor. purity: 99%