Learn More
Medchemexpress LLC Avitinib maleate | 1557268-88-8 | 99.86% | 603.60 | 1 ML
SDP
Catalog No. 5000400850
Encompass_Preferred
Avitinib maleate (Abivertinib) is a third-generation, irreversible, and orally active selective EGFR inhibitor. It has IC50 values of 0.18 nM against EGFR L858R and EGFR T790M, and 7.68 nM against wild-type EGFR. Additionally, Avitinib maleate acts as a BTK inhibitor, inducing apoptosis and inhibiting BTK phosphorylation in mantle cell lymphoma. This compound demonstrates anticancer effects and is intended for research use only.
  • Third-generation, irreversible, and orally active selective EGFR inhibitor.
  • Potent inhibition of mutant EGFR phosphorylation with high selectivity over wild-type EGFR.
  • Inhibits phosphorylation of downstream targets such as Akt and ERK1/2 in specific cell lines.
  • Also functions as a BTK inhibitor, inducing apoptosis and inhibiting BTK phosphorylation in mantle cell lymphoma.
  • Demonstrates anticancer effects in xenograft models, inhibiting EGFR-mutant tumor growth.
Catalog No. 50-004-00850 Supplier Medchemexpress LLC Supplier No. HY19816A10MM/1ML
Avitinib maleate (Abivertinib) is a third-generation, irreversible, and orally active selective EGFR inhibitor. It has IC50 values of 0.18 nM against EGFR L858R and EGFR T790M, and 7.68 nM against wild-type EGFR. Additionally, Avitinib maleate acts as a BTK inhibitor, inducing apoptosis and inhibiting BTK phosphorylation in mantle cell lymphoma. This compound demonstrates anticancer effects and is intended for research use only.
  • Third-generation, irreversible, and orally active selective EGFR inhibitor.
  • Potent inhibition of mutant EGFR phosphorylation with high selectivity over wild-type EGFR.
  • Inhibits phosphorylation of downstream targets such as Akt and ERK1/2 in specific cell lines.
  • Also functions as a BTK inhibitor, inducing apoptosis and inhibiting BTK phosphorylation in mantle cell lymphoma.
  • Demonstrates anticancer effects in xenograft models, inhibiting EGFR-mutant tumor growth.