Learn More
Medchemexpress LLC Avitinib maleate | 1557268-88-8 | 99.9% | 603.60 | 100 MG
SDP
Catalog No. 5000400849
Encompass_Preferred
Avitinib (Abivertinib) maleate is a third-generation, irreversible and orally active selective EGFR inhibitor, with IC50 values of 0.18 nM, 0.18 nM, 7.68 nM and against EGFR L858R, EGFR T790M and wild-type EGFR. Avitinib maleate is also a BTK inhibitor that induces apoptosis and inhibits phosphorylation of BTK in mantle cell lymphoma. Avitinib maleate shows anticancer effects.
  • Third-generation, irreversible and orally active selective EGFR inhibitor.
  • Potent inhibition of EGFR L858R (IC50: 0.18 nM), EGFR T790M (IC50: 0.18 nM), and wild-type EGFR (IC50: 7.68 nM).
  • BTK inhibitor that induces apoptosis and inhibits phosphorylation of BTK in mantle cell lymphoma.
  • Exhibits anticancer effects.
  • Selectively inhibits mutant EGFR phosphorylation.
  • Inhibits phosphorylation of downstream targets Akt and ERK1/2 in NCI-H1975 and HCC827 cells.
  • Inhibits EGFR-mutant tumor growth in xenograft models over extended duration.
Catalog No. 50-004-00849 Supplier Medchemexpress LLC Supplier No. HY19816A100MG
Avitinib (Abivertinib) maleate is a third-generation, irreversible and orally active selective EGFR inhibitor, with IC50 values of 0.18 nM, 0.18 nM, 7.68 nM and against EGFR L858R, EGFR T790M and wild-type EGFR. Avitinib maleate is also a BTK inhibitor that induces apoptosis and inhibits phosphorylation of BTK in mantle cell lymphoma. Avitinib maleate shows anticancer effects.
  • Third-generation, irreversible and orally active selective EGFR inhibitor.
  • Potent inhibition of EGFR L858R (IC50: 0.18 nM), EGFR T790M (IC50: 0.18 nM), and wild-type EGFR (IC50: 7.68 nM).
  • BTK inhibitor that induces apoptosis and inhibits phosphorylation of BTK in mantle cell lymphoma.
  • Exhibits anticancer effects.
  • Selectively inhibits mutant EGFR phosphorylation.
  • Inhibits phosphorylation of downstream targets Akt and ERK1/2 in NCI-H1975 and HCC827 cells.
  • Inhibits EGFR-mutant tumor growth in xenograft models over extended duration.