Learn More
Medchemexpress LLC Avitinib maleate | 1557268-88-8 | 99.9% | 603.60 | 25 MG
SDP
Catalog No. 5000402668
Encompass_Preferred
Avitinib maleate is a third-generation, irreversible, and orally active selective EGFR inhibitor. It has IC50 values of 0.18 nM against both EGFR L858R and EGFR T790M, and 7.68 nM against wild-type EGFR. It is also a BTK inhibitor, inducing apoptosis and inhibiting BTK phosphorylation in mantle cell lymphoma. It exhibits anticancer effects.
  • Third-generation, irreversible, and orally active selective EGFR inhibitor.
  • Potent against EGFR L858R, EGFR T790M, and wild-type EGFR.
  • BTK inhibitor.
  • Induces apoptosis.
  • Inhibits phosphorylation of BTK in mantle cell lymphoma.
  • Exhibits anticancer effects.
Catalog No. 50-004-02668 Supplier Medchemexpress LLC Supplier No. 25MGHY19816A
Avitinib maleate is a third-generation, irreversible, and orally active selective EGFR inhibitor. It has IC50 values of 0.18 nM against both EGFR L858R and EGFR T790M, and 7.68 nM against wild-type EGFR. It is also a BTK inhibitor, inducing apoptosis and inhibiting BTK phosphorylation in mantle cell lymphoma. It exhibits anticancer effects.
  • Third-generation, irreversible, and orally active selective EGFR inhibitor.
  • Potent against EGFR L858R, EGFR T790M, and wild-type EGFR.
  • BTK inhibitor.
  • Induces apoptosis.
  • Inhibits phosphorylation of BTK in mantle cell lymphoma.
  • Exhibits anticancer effects.