Learn More
Medchemexpress LLC Avitinib maleate | 1557268-88-8 | 99.9% | 603.60 | 50 MG
SDP
Catalog No. 5000401178
Encompass_Preferred
Avitinib maleate (Abivertinib maleate) is a third-generation, irreversible and orally active selective EGFR inhibitor. It also functions as a BTK inhibitor that induces apoptosis and inhibits phosphorylation of BTK in mantle cell lymphoma. It demonstrates anticancer effects.
  • Third-generation, irreversible, and orally active selective EGFR inhibitor.
  • Potent against EGFR L858R, EGFR T790M, and wild-type EGFR.
  • BTK inhibitor.
  • Induces apoptosis.
  • Inhibits phosphorylation of BTK in mantle cell lymphoma.
  • Shows anticancer effects.
Catalog No. 50-004-01178 Supplier Medchemexpress LLC Supplier No. HY19816A50MG
Avitinib maleate (Abivertinib maleate) is a third-generation, irreversible and orally active selective EGFR inhibitor. It also functions as a BTK inhibitor that induces apoptosis and inhibits phosphorylation of BTK in mantle cell lymphoma. It demonstrates anticancer effects.
  • Third-generation, irreversible, and orally active selective EGFR inhibitor.
  • Potent against EGFR L858R, EGFR T790M, and wild-type EGFR.
  • BTK inhibitor.
  • Induces apoptosis.
  • Inhibits phosphorylation of BTK in mantle cell lymphoma.
  • Shows anticancer effects.