Learn More
Medchemexpress LLC Avitinib maleate | 1557268-88-8 | MFCD29089376 | 99.9% | 603.60 | C30H30FN7O6 | 10 MG
SDP
Catalog No. 5000297700
Encompass_Preferred
Avitinib maleate is a third-generation, irreversible, orally active EGFR inhibitor supplied as the maleate salt for research use. It exhibits potent activity against EGFR L858R and T790M mutations, with reported BTK inhibitory activity, and is provided with analytical data confirming high purity. Typical formats include small solid quantities and DMSO solutions for laboratory studies.
  • Third-generation, irreversible EGFR inhibitor suitable for targeted research.
  • Potent activity against EGFR L858R and T790M mutants.
  • Reduced potency against wild-type EGFR to aid selectivity studies.
  • Maleate salt form for improved handling and stability.
  • High purity with RP-HPLC documentation.
  • Available in small solid pack sizes and solution formats for assays.
  • Intended for in vitro and preclinical research applications.
Catalog No. 50-002-97700 Supplier Medchemexpress LLC Supplier No. HY19816A10MG
Avitinib maleate is a third-generation, irreversible, orally active EGFR inhibitor supplied as the maleate salt for research use. It exhibits potent activity against EGFR L858R and T790M mutations, with reported BTK inhibitory activity, and is provided with analytical data confirming high purity. Typical formats include small solid quantities and DMSO solutions for laboratory studies.
  • Third-generation, irreversible EGFR inhibitor suitable for targeted research.
  • Potent activity against EGFR L858R and T790M mutants.
  • Reduced potency against wild-type EGFR to aid selectivity studies.
  • Maleate salt form for improved handling and stability.
  • High purity with RP-HPLC documentation.
  • Available in small solid pack sizes and solution formats for assays.
  • Intended for in vitro and preclinical research applications.