Learn More
Medchemexpress LLC Avj16 | 2775241-92-2 | 99.6% | C28H27N3O4 | 10MG
SDP
Catalog No. 5000258176
Encompass_Preferred
AVJ16 is a small-molecule inhibitor of IGF2BP1 used in research to inhibit cancer cell migration and to study IGF2BP1-mediated regulation of mRNA translation. Reported affinity is Kd = 1.4 μM; the compound has been characterized for effects on proliferation, invasion, and migration in IGF2BP1-expressing cancer cell lines. Molecular formula C28H27N3O4, molecular weight 469.53.
Catalog No. 50-002-58176 Supplier Medchemexpress LLC Supplier No. HY14487310MG
AVJ16 is a small-molecule inhibitor of IGF2BP1 used in research to inhibit cancer cell migration and to study IGF2BP1-mediated regulation of mRNA translation. Reported affinity is Kd = 1.4 μM; the compound has been characterized for effects on proliferation, invasion, and migration in IGF2BP1-expressing cancer cell lines. Molecular formula C28H27N3O4, molecular weight 469.53.