Learn More
Apexbio Technology LLC AVL-292, 10mg. Cas: 1202757-89-8 MFCD: MFCD25976876. Btk inhibitor
SDP
Supplier:  Apexbio Technology LLC A320610
Encompass_Preferred
CC-292, formerly known as AVL-292, is an orally active, potent irreversible small molecule inhibitor of BTK with IC50 of 0.5 nM and EC50 of 8 nM. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4788