Learn More
Apexbio Technology LLC AVL-292 1202757-89-8 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A32065.1
Encompass_Preferred
AVL-292 (CAS 1202757-89-8) also known as CC-292 is an orally bioavailable irreversible small-molecule inhibitor targeting Bruton s tyrosine kinase (BTK) a member of the Tec kinase family crucial for B-cell receptor (BCR) signaling pathways AVL-292 covalently binds the cysteine residue (Cys481) within BTK suppressing its autophosphorylation and downstream signaling Reported IC50 and EC50 values in biochemical and cellular assays are 0 5 nM and 8 nM respectively AVL-292 has shown efficacy in preclinical models such as collagen-induced arthritis (CIA) mice and demonstrates therapeutic potential in clinical studies involving B-cell malignancies including CLL B-NHL and WM
Catalog No. 50-000-62934