Learn More
Medchemexpress LLC AVN-101 hydrochlorid 100mg | 1061354-48-0 | 100 MG
SDP
Catalog No. 5000308352
Encompass_Preferred
AVN-101 hydrochloride is a brain-penetrant, orally active small-molecule antagonist of the 5-HT7 serotonin receptor with additional affinity for 5-HT6, 5-HT2A/2C, histamine H1, and adrenergic α2 receptors. Supplied as a high-purity solid for preclinical CNS research.
  • Potent 5-HT7 antagonist (Ki ≈ 153 pM) with multi-receptor activity.
  • Brain-penetrant and orally active, suitable for in vivo studies.
  • High purity (99.59%) solid suitable for analytical and biological assays.
  • Molecular weight 340.9 g/mol; formula C21H25ClN2.
  • Available in multiple sizes, including a 100 mg pack for small-scale studies.
Catalog No. 50-003-08352 Supplier Medchemexpress LLC Supplier No. HY117046A100MG
AVN-101 hydrochloride is a brain-penetrant, orally active small-molecule antagonist of the 5-HT7 serotonin receptor with additional affinity for 5-HT6, 5-HT2A/2C, histamine H1, and adrenergic α2 receptors. Supplied as a high-purity solid for preclinical CNS research.
  • Potent 5-HT7 antagonist (Ki ≈ 153 pM) with multi-receptor activity.
  • Brain-penetrant and orally active, suitable for in vivo studies.
  • High purity (99.59%) solid suitable for analytical and biological assays.
  • Molecular weight 340.9 g/mol; formula C21H25ClN2.
  • Available in multiple sizes, including a 100 mg pack for small-scale studies.