Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AVN-101 hydrochlorid 10mg | 1061354-48-0 | 340.89 | C21H25ClN2 | 10 MG
SDP

Catalog No. 5000308326
Encompass_Preferred

AVN-101 hydrochloride is the hydrochloride salt of AVN-101, a brain-penetrant, orally active antagonist of serotonin 5-HT7 receptors used in preclinical CNS research. It is supplied as a high-purity solid for in vitro and in vivo studies.

  • Potent 5-HT7 receptor antagonist with additional activity at 5-HT6, 5-HT2A, 5-HT2C, H1, and α2 adrenergic receptors.
  • Brain-penetrant and orally active, suitable for CNS pharmacology studies.
  • High purity solid (LCMS purity 99.59%) for reliable experimental results.
  • Chemical identity: 1H-pyrido[4,3-b]indole hydrochloride; formula C21H25ClN2; Mw 340.89.
  • Recommended storage: sealed at -20°C; solvent storage guidance available from supplier.

Catalog No. 50-003-08326 Supplier Medchemexpress LLC Supplier No. HY117046A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AVN-101 hydrochloride is the hydrochloride salt of AVN-101, a brain-penetrant, orally active antagonist of serotonin 5-HT7 receptors used in preclinical CNS research. It is supplied as a high-purity solid for in vitro and in vivo studies.

  • Potent 5-HT7 receptor antagonist with additional activity at 5-HT6, 5-HT2A, 5-HT2C, H1, and α2 adrenergic receptors.
  • Brain-penetrant and orally active, suitable for CNS pharmacology studies.
  • High purity solid (LCMS purity 99.59%) for reliable experimental results.
  • Chemical identity: 1H-pyrido[4,3-b]indole hydrochloride; formula C21H25ClN2; Mw 340.89.
  • Recommended storage: sealed at -20°C; solvent storage guidance available from supplier.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.