Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AVN-101 hydrochlorid 25mg | 1061354-48-0 | 340.89 | C21H25ClN2 | 25 MG
SDP

Catalog No. 5000229275
Encompass_Preferred

AVN-101 hydrochloride is the hydrochloride salt of a brain-penetrant small molecule antagonist of serotonin (5-HT) receptors, supplied for preclinical and in-vitro research. It is provided as a solid with reported high purity for analytical and biological assays.

  • Potent antagonist of 5-HT receptor subtypes (5-HT7, 5-HT6, 5-HT2A, 5-HT2C).
  • High purity: 99.59%.
  • Molecular formula C21H25ClN2; molecular weight 340.89.
  • Available in small research pack sizes, including 25 MG.
  • Provided as hydrochloride salt to improve solubility for assay preparation.
  • For research use only; not for human or veterinary use.

Catalog No. 50-002-29275 Supplier Medchemexpress LLC Supplier No. HY117046A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AVN-101 hydrochloride is the hydrochloride salt of a brain-penetrant small molecule antagonist of serotonin (5-HT) receptors, supplied for preclinical and in-vitro research. It is provided as a solid with reported high purity for analytical and biological assays.

  • Potent antagonist of 5-HT receptor subtypes (5-HT7, 5-HT6, 5-HT2A, 5-HT2C).
  • High purity: 99.59%.
  • Molecular formula C21H25ClN2; molecular weight 340.89.
  • Available in small research pack sizes, including 25 MG.
  • Provided as hydrochloride salt to improve solubility for assay preparation.
  • For research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.