Learn More
Medchemexpress LLC AVN-101 hydrochlorid 5mg | 1061354-48-0 | 340.89 g/mol | C21H25ClN2 | 5 MG
SDP
Catalog No. 5000308344
Encompass_Preferred
AVN-101 hydrochloride is a brain-penetrant, orally active 5-HT7 receptor antagonist used in pharmacological research. Supplied as a 5 mg powder with high purity, it is suitable for in vitro assays and preclinical studies requiring selective serotonergic modulation.
  • High purity suitable for research applications.
  • Brain-penetrant profile for central nervous system studies.
  • Orally active salt form compatible with dosing studies.
  • Stable powder for sealed, low-temperature storage.
  • Small packaging appropriate for early-stage research.
Catalog No. 50-003-08344 Supplier Medchemexpress LLC Supplier No. HY117046A5MG
AVN-101 hydrochloride is a brain-penetrant, orally active 5-HT7 receptor antagonist used in pharmacological research. Supplied as a 5 mg powder with high purity, it is suitable for in vitro assays and preclinical studies requiring selective serotonergic modulation.
  • High purity suitable for research applications.
  • Brain-penetrant profile for central nervous system studies.
  • Orally active salt form compatible with dosing studies.
  • Stable powder for sealed, low-temperature storage.
  • Small packaging appropriate for early-stage research.