Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Avn-492 | 1220646-23-0 | 99.3% | 359.45 g/mol | C17H21N5O2S | 1 ML
SDP

Catalog No. 5000246514
Encompass_Preferred

AVN-492 is a selective, high-affinity antagonist of the serotonin 5-HT6 receptor (Ki ≈ 91 pM) used as a research tool in pharmacology and neuroscience to investigate 5-HT6-mediated signaling, cognition, and neurodegenerative disease mechanisms. It is supplied with high purity and is available as both solid material and a DMSO solution for in vitro assays.

  • High affinity 5-HT6 receptor antagonist (Ki ≈ 91 pM)
  • High purity suitable for research applications (99.3%)
  • Available as solid and 10 mM DMSO solution for flexibility in assays
  • Soluble in DMSO (reported 50 mg/mL) for easy stock preparation
  • Recommended storage conditions for stability: powder -20°C; in solvent -80°C

Catalog No. 50-002-46514 Supplier Medchemexpress LLC Supplier No. HY10192410MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AVN-492 is a selective, high-affinity antagonist of the serotonin 5-HT6 receptor (Ki ≈ 91 pM) used as a research tool in pharmacology and neuroscience to investigate 5-HT6-mediated signaling, cognition, and neurodegenerative disease mechanisms. It is supplied with high purity and is available as both solid material and a DMSO solution for in vitro assays.

  • High affinity 5-HT6 receptor antagonist (Ki ≈ 91 pM)
  • High purity suitable for research applications (99.3%)
  • Available as solid and 10 mM DMSO solution for flexibility in assays
  • Soluble in DMSO (reported 50 mg/mL) for easy stock preparation
  • Recommended storage conditions for stability: powder -20°C; in solvent -80°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.