Learn More
Medchemexpress LLC Avn-492 | 1220646-23-0 | MFCD31544338 | 99.3% | 359.45 g·mol⁻¹ | C17H21N5O2S | 10 MG
SDP
Catalog No. 5000246961
Encompass_Preferred
AVN-492 is a potent, highly selective antagonist of the serotonin 5-HT6 receptor intended for research use. It exhibits picomolar affinity (reported Ki ≈ 91 pM) and is supplied as a solid powder for in vitro and in vivo pharmacology, receptor binding, and functional assays. Recommended storage and quality details are provided on the supplier certificate of analysis.
  • Potent 5-HT6 receptor antagonist with picomolar affinity (Ki ≈ 91 pM).
  • High chemical purity (≈99.3%).
  • Supplied as a solid powder for flexible use in assays.
  • Suitable for receptor binding, functional pharmacology, and in vivo studies.
  • Stable when stored under recommended conditions.
  • Molecular formula C17H21N5O2S and molecular weight 359.45 g·mol⁻¹.
Catalog No. 50-002-46961 Supplier Medchemexpress LLC Supplier No. HY10192410MG
AVN-492 is a potent, highly selective antagonist of the serotonin 5-HT6 receptor intended for research use. It exhibits picomolar affinity (reported Ki ≈ 91 pM) and is supplied as a solid powder for in vitro and in vivo pharmacology, receptor binding, and functional assays. Recommended storage and quality details are provided on the supplier certificate of analysis.
  • Potent 5-HT6 receptor antagonist with picomolar affinity (Ki ≈ 91 pM).
  • High chemical purity (≈99.3%).
  • Supplied as a solid powder for flexible use in assays.
  • Suitable for receptor binding, functional pharmacology, and in vivo studies.
  • Stable when stored under recommended conditions.
  • Molecular formula C17H21N5O2S and molecular weight 359.45 g·mol⁻¹.