Learn More
Medchemexpress LLC HY-101924 5mg Medchemexpress, AVN-492 CAS:1220646-23-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1019245MG
Encompass_Preferred
Medchemexpress, HY-101924 5mg AVN-492 CAS:1220646-23-0 AVN-492 is a very specific and highly-selective antagonist with picomolar affinity to 5-HT6R (Ki=91 pM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7918