Learn More
Cayman Chemical AvnafIl-13C-d3 1mg
SDP
Catalog No. 502988073
Encompass_Preferred
An internal standard for the quantification of avanafil by GC- or LC-MS
Catalog No. 50-298-8073 Supplier Cayman Chemical Supplier No. 316871 MG
An internal standard for the quantification of avanafil by GC- or LC-MS