Learn More
Cayman Chemical Avobenzone-13C-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 2933758-47-3, 1 mg
SDP
Catalog No. 502947055
Encompass_Preferred
An internal standard for the quantification of avobenzone by GC- or LC-MS
Catalog No. 50-294-7055 Supplier Cayman Chemical Supplier No. 333671 MG
An internal standard for the quantification of avobenzone by GC- or LC-MS