Learn More
Cayman Chemical Avobenzon-13C-d3 5mg
SDP
Supplier:  Cayman Chemical 333675 MG
Encompass_Preferred
An internal standard for the quantification of avobenzone by GC- or LC-MS
Catalog No. 50-294-8106